Learn More
Medchemexpress LLC HY-135829 5mg Medchemexpress, BAY 2416964 CAS:2242464-44-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1358295MG
Encompass_Preferred
Medchemexpress, HY-135829 5mg BAY 2416964 CAS:2242464-44-2 BAY 2416964 is a potent and orally active aryl hydrocarbon receptor (AHR) antagonist extracted from patent WO2018146010A1, example 192, has an IC50 of 341 nM. BAY 2416964 has the potential for solid tumors treatment. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0366