Learn More
Selleck Chemical LLC BAY 2416964 S8995-10MM/1ML
SDP
Catalog No. 5000025010
Encompass_Preferred
BAY 2416964 (compound 192) is a potent and orally active antagonist of aryl hydrocarbon receptor (AhR) with IC50 of 341 nM BAY 2416964 has the potential in the treatment of solid tumors
Catalog No. 50-000-25010 Supplier Selleck Chemical LLC Supplier No. S899510MM/1ML
BAY 2416964 (compound 192) is a potent and orally active antagonist of aryl hydrocarbon receptor (AhR) with IC50 of 341 nM BAY 2416964 has the potential in the treatment of solid tumors