Learn More
Medchemexpress LLC BAY 249716 | 696628-24-7 | 99.0% | 288.76 | 5 MG
SDP
Catalog No. 5000394859
Encompass_Preferred
BAY 249716 stabilizes all three p53 protein variants. It also exhibits antitubercular activity with an IC90 of <0.10 μg/mL for Tuberculosis. This product is for research use only and not for sale to patients.
  • Stabilizes all three p53 protein variants.
  • Exhibits antitubercular activity with an IC90 of <0.10 μg/mL for Tuberculosis.
  • Antileishmanial activity against Leishmania donovani in mouse J-774A1 cells (IC50: 0.09 μM).
Catalog No. 50-003-94859 Supplier Medchemexpress LLC Supplier No. HY1609225MG
BAY 249716 stabilizes all three p53 protein variants. It also exhibits antitubercular activity with an IC90 of <0.10 μg/mL for Tuberculosis. This product is for research use only and not for sale to patients.
  • Stabilizes all three p53 protein variants.
  • Exhibits antitubercular activity with an IC90 of <0.10 μg/mL for Tuberculosis.
  • Antileishmanial activity against Leishmania donovani in mouse J-774A1 cells (IC50: 0.09 μM).