Learn More
Medchemexpress LLC BAY-293 10mg | 2244904-70-7 | 10 MG

Supplier: Medchemexpress LLC HY11439810MG
BAY-293 is a small-molecule chemical probe that inhibits the SOS1-KRAS interaction, reducing RAS activation (reported IC50 ≈ 21 nM). Supplied as a purified solid for research use; store at -20°C.
- Potent SOS1-KRAS interaction inhibitor (IC50 ≈ 21 nM).
- High purity, ≥98% by HPLC.
- Molecular weight 448.59 g·mol⁻1.
- Chemical formula C25H28N4O2S.
- Supplied as a milligram-scale solid; store at -20°C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.