Learn More
Medchemexpress LLC HY-114398 50mg , BAY-293 CAS:2244904-70-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11439850MG
Encompass_Preferred
HY-114398 50mg BAY-293 CAS: 2244904-70-7 BAY-293, a valuable chemical probe, blocks RAS activation via disruption of the KRAS-SOS1 interaction with an IC50 of 21 nM. BAY-293 is a potent inhibitor of Son of Sevenless 1 (SOS1). SOS1 is the guanine nucleotide exchange factor (GEF) and activator of RAS. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0567