Learn More
Medchemexpress LLC BAY-298 | 2471978-97-7 | 100.0% | 100 MG
SDP
Catalog No. 5000340061
Encompass_Preferred
BAY-298 is an orally active and selective luteinizing hormone receptor (LH-R) antagonist. It has IC50s of 96 nM for human LH (hLH), 23 nM for rat LH (rLH), and 78 nM for cynomolgus monkey LH (cLH). This compound is capable of reducing sex hormone levels and is intended for research use only.
  • Orally active and selective luteinizing hormone receptor (LH-R) antagonist
  • Capable of reducing sex hormone levels
  • IC50 of 96 nM for human LH, 23 nM for rat LH, and 78 nM for cynomolgus monkey LH
  • Solid appearance
  • Off-white to light yellow color
Catalog No. 50-003-40061 Supplier Medchemexpress LLC Supplier No. HY130249100MG
BAY-298 is an orally active and selective luteinizing hormone receptor (LH-R) antagonist. It has IC50s of 96 nM for human LH (hLH), 23 nM for rat LH (rLH), and 78 nM for cynomolgus monkey LH (cLH). This compound is capable of reducing sex hormone levels and is intended for research use only.
  • Orally active and selective luteinizing hormone receptor (LH-R) antagonist
  • Capable of reducing sex hormone levels
  • IC50 of 96 nM for human LH, 23 nM for rat LH, and 78 nM for cynomolgus monkey LH
  • Solid appearance
  • Off-white to light yellow color