Learn More
Medchemexpress LLC BAY-298 | 2471978-97-7 | 100.0% | 25 MG
SDP
Catalog No. 5000342207
Encompass_Preferred
BAY-298 is an orally active and selective luteinizing hormone receptor (LH-R) antagonist that can reduce sex hormone levels, making it suitable for research applications related to metabolic or endocrine diseases.
  • Orally active and selective luteinizing hormone receptor (LH-R) antagonist.
  • Reduces sex hormone levels.
  • Inhibits human ERG in HEK293 cells (IC50: 3 μM).
  • Exhibits antagonist activity at cynomolgus monkey luteinizing hormone receptor in HEK293 cells (IC50: 78 nM).
  • Lowers serum estradiol levels in intact female rats.
Catalog No. 50-003-42207 Supplier Medchemexpress LLC Supplier No. 25MGHY130249
BAY-298 is an orally active and selective luteinizing hormone receptor (LH-R) antagonist that can reduce sex hormone levels, making it suitable for research applications related to metabolic or endocrine diseases.
  • Orally active and selective luteinizing hormone receptor (LH-R) antagonist.
  • Reduces sex hormone levels.
  • Inhibits human ERG in HEK293 cells (IC50: 3 μM).
  • Exhibits antagonist activity at cynomolgus monkey luteinizing hormone receptor in HEK293 cells (IC50: 78 nM).
  • Lowers serum estradiol levels in intact female rats.