Learn More
Medchemexpress LLC BAY-298 | 2471978-97-7 | 99.9% | 10 MM * 1 ML
SDP
Catalog No. 5000341045
Encompass_Preferred
BAY-298 is an orally active and selective luteinizing hormone receptor (LH-R) antagonist. It demonstrates potent activity against human, rat, and cynomolgus monkey LH receptors, effectively reducing sex hormone levels. It is supplied as a 10 MM solution in DMSO.
  • Orally active and selective LH-R antagonist
  • Reduces sex hormone levels
  • Potent inhibitory activity against human, rat, and cynomolgus monkey LH receptors
  • Inhibits human ERG in HEK293 cells
  • Lowers serum estradiol levels in proestrus female rats
Catalog No. 50-003-41045 Supplier Medchemexpress LLC Supplier No. HY13024910MM/1ML
BAY-298 is an orally active and selective luteinizing hormone receptor (LH-R) antagonist. It demonstrates potent activity against human, rat, and cynomolgus monkey LH receptors, effectively reducing sex hormone levels. It is supplied as a 10 MM solution in DMSO.
  • Orally active and selective LH-R antagonist
  • Reduces sex hormone levels
  • Potent inhibitory activity against human, rat, and cynomolgus monkey LH receptors
  • Inhibits human ERG in HEK293 cells
  • Lowers serum estradiol levels in proestrus female rats