Learn More
Medchemexpress LLC HY-107424 10mg , BAY-299 CAS:2080306-23-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10742410MG
Encompass_Preferred
HY-107424 10mg BAY-299 CAS: 2080306-23-4 BAY-299 is a very potent, dual inhibitor with IC50s of 67 nM for BRPF2 bromodomains (BD), 8 nM for TAF1 BD2, and 106 nM for TAF1L BD2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0486