Learn More
Medchemexpress LLC Bay-299 | 2080306-23-4 | MFCD30480933 | >98.0% | 429.47 Da | C25H23N3O4 | 25 MG
SDP
Catalog No. 5000239735
Encompass_Preferred
BAY-299 is a small-molecule chemical probe that potently inhibits bromodomain-containing proteins BRPF2/BRD1 and the second bromodomain of TAF1/TAF1L. It is for research use only and has been reported with low-nanomolar biochemical potency and cellular activity in published and vendor assays.
  • Potent dual bromodomain inhibitor of BRPF2/BRD1 and TAF1.
  • Low-nanomolar biochemical potency in reported assays.
  • Selective over many other bromodomains, reducing off-target binding.
  • Demonstrated cell activity in NanoBRET and related assays.
  • Supplied as a high-purity solid suitable for research applications.
  • Useful for mechanistic studies of chromatin and transcriptional regulation.
Catalog No. 50-002-39735 Supplier Medchemexpress LLC Supplier No. HY10742425MG
BAY-299 is a small-molecule chemical probe that potently inhibits bromodomain-containing proteins BRPF2/BRD1 and the second bromodomain of TAF1/TAF1L. It is for research use only and has been reported with low-nanomolar biochemical potency and cellular activity in published and vendor assays.
  • Potent dual bromodomain inhibitor of BRPF2/BRD1 and TAF1.
  • Low-nanomolar biochemical potency in reported assays.
  • Selective over many other bromodomains, reducing off-target binding.
  • Demonstrated cell activity in NanoBRET and related assays.
  • Supplied as a high-purity solid suitable for research applications.
  • Useful for mechanistic studies of chromatin and transcriptional regulation.