Learn More
TARGETMOL CHEMICALS INC BAY-299 5MG
SDP
Catalog No. 502581285
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BAY-299 is an effecitve inhibitor of the bromodomain and PHD finger family member BRPF2 and the TATA box binding protein-associated factors TAF1 and TAF1L with IC50s of 67 nM 8 nM and 106 nM respectively. purity: 97%
Catalog No. 50-258-1285 Supplier TARGETMOL CHEMICALS INC Supplier No. T145025MG
Also available in 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BAY-299 is an effecitve inhibitor of the bromodomain and PHD finger family member BRPF2 and the TATA box binding protein-associated factors TAF1 and TAF1L with IC50s of 67 nM 8 nM and 106 nM respectively. purity: 97%