Learn More
Abcam BAY-299, TAF1 and BRD1 inhibitor, 10MG
Catalog No. 502676153
Encompass_Preferred
MW 429.5 Da, Purity >98%. Potent and selective bromodomain inhibitor of TAF1 (IC₅₀ = 8-13 nM) and BRD1 (IC₅₀ = 6-67 nM). More than 30-fold selective versus BRD9 and ATAD2. More than 300-fold selective versus BRD4. Inhibits BRD1 (IC₅₀ = 0.58 μM) and TAF1 (IC₅₀ = 0.9 μM) binding to Histone H4 in cell-based assays; also inhibits BRD1 (IC₅₀ = 0.83 μM) and TAF1 (IC₅₀ = 1.4 μM) binding to Histone H3.3.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-6153 Supplier Abcam Supplier No. AB23036810MG
MW 429.5 Da, Purity >98%. Potent and selective bromodomain inhibitor of TAF1 (IC₅₀ = 8-13 nM) and BRD1 (IC₅₀ = 6-67 nM). More than 30-fold selective versus BRD9 and ATAD2. More than 300-fold selective versus BRD4. Inhibits BRD1 (IC₅₀ = 0.58 μM) and TAF1 (IC₅₀ = 0.9 μM) binding to Histone H4 in cell-based assays; also inhibits BRD1 (IC₅₀ = 0.83 μM) and TAF1 (IC₅₀ = 1.4 μM) binding to Histone H3.3.
The product is subject to the following: Abcam Restricted Use Statement