Learn More
Medchemexpress LLC BAY-320 | 1445830-50-1 | MFCD31657346 | 99.7% | 492.52 g/mol | C26H26F2N6O2 | 10 MG
SDP
Catalog No. 5000243498
Encompass_Preferred
BAY-320 is a small-molecule, ATP-competitive inhibitor of the Bub1 serine/threonine kinase used as a research tool to probe Bub1 catalytic function, mitotic progression, and sensitivity to microtubule-targeting agents. It demonstrates selective biochemical and cellular activity and is supplied for research use as a solid or as a 10 mM solution in DMSO.
  • Selective ATP-competitive Bub1 kinase inhibitor.
  • Documented biochemical and cellular activity at low micromolar concentrations.
  • Useful for studying mitosis, chromosome segregation, and drug sensitization.
  • Available as a solid and as a 10 mM solution in DMSO for convenience.
  • High purity suitable for research applications.
  • Stable when stored at recommended cold temperatures.
Catalog No. 50-002-43498 Supplier Medchemexpress LLC Supplier No. HY10400010MG
BAY-320 is a small-molecule, ATP-competitive inhibitor of the Bub1 serine/threonine kinase used as a research tool to probe Bub1 catalytic function, mitotic progression, and sensitivity to microtubule-targeting agents. It demonstrates selective biochemical and cellular activity and is supplied for research use as a solid or as a 10 mM solution in DMSO.
  • Selective ATP-competitive Bub1 kinase inhibitor.
  • Documented biochemical and cellular activity at low micromolar concentrations.
  • Useful for studying mitosis, chromosome segregation, and drug sensitization.
  • Available as a solid and as a 10 mM solution in DMSO for convenience.
  • High purity suitable for research applications.
  • Stable when stored at recommended cold temperatures.