Learn More
Medchemexpress LLC Bay-320 | 1445830-50-1 | MFCD31657346 | 99.7% | 492.52 g/mol | C26H26F2N6O2 | 50 MG
SDP
Catalog No. 5000243215
Encompass_Preferred
BAY-320 is a selective, ATP-competitive inhibitor of the Bub1 kinase used in biochemical and cellular research. It inhibits Bub1 catalytic activity (IC50 ≈ 680 nM), disrupts Bub1-mediated Sgo1 localization, alters mitotic chromosome arrangement and spindle assembly, and reduces cell proliferation, making it useful in cancer research applications such as studies of ovarian and cervical cancer.
  • Selective ATP-competitive Bub1 kinase inhibitor
  • Biochemical and cellular activity with reported IC50 ≈ 680 nM
  • Disrupts Sgo1 localization and mitotic chromosome arrangement
  • Available as powder and as a 10 mM solution in DMSO for flexible use
  • High purity suitable for research applications
Catalog No. 50-002-43215 Supplier Medchemexpress LLC Supplier No. HY10400050MG
BAY-320 is a selective, ATP-competitive inhibitor of the Bub1 kinase used in biochemical and cellular research. It inhibits Bub1 catalytic activity (IC50 ≈ 680 nM), disrupts Bub1-mediated Sgo1 localization, alters mitotic chromosome arrangement and spindle assembly, and reduces cell proliferation, making it useful in cancer research applications such as studies of ovarian and cervical cancer.
  • Selective ATP-competitive Bub1 kinase inhibitor
  • Biochemical and cellular activity with reported IC50 ≈ 680 nM
  • Disrupts Sgo1 localization and mitotic chromosome arrangement
  • Available as powder and as a 10 mM solution in DMSO for flexible use
  • High purity suitable for research applications