Learn More
Medchemexpress LLC HY-104000 5mg , BAY-320 CAS:1445830-50-1 Purity:>98%
SDP
Catalog No. 502261629
Encompass_Preferred
HY-104000 5mg BAY-320 CAS: 1445830-50-1 BAY-320 is a Bub1 inhibitor, with an IC50 of 680 nM for human Bub1 in the presence of 2 mM ATP. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-1629 Supplier Medchemexpress LLC Supplier No. HY1040005MG
HY-104000 5mg BAY-320 CAS: 1445830-50-1 BAY-320 is a Bub1 inhibitor, with an IC50 of 680 nM for human Bub1 in the presence of 2 mM ATP. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.