Learn More
Selleck Chemical LLC BAY-3827-5mg
SDP
Catalog No. 502389877
Encompass_Preferred
BAY-3827 is a potent AMPK inhibitor which inhibits AMPK kinase activity with IC50 values of 1.4 nM at a low, 10 μM ATP concentration and 15 nM at a high, 2 mM ATP concentration.
Catalog No. 50-238-9877 Supplier Selleck Chemical LLC Supplier No. S98335MG
BAY-3827 is a potent AMPK inhibitor which inhibits AMPK kinase activity with IC50 values of 1.4 nM at a low, 10 μM ATP concentration and 15 nM at a high, 2 mM ATP concentration.