Learn More
Selleck Chemical LLC BAY 41-2272
SDP
Catalog No. 5000001391
Encompass_Preferred
BAY 41-2272 is a direct and NO-independent soluble guanylate cyclase (sGC) stimulator
Catalog No. 50-000-01391 Supplier Selleck Chemical LLC Supplier No. S812825MG
BAY 41-2272 is a direct and NO-independent soluble guanylate cyclase (sGC) stimulator