Learn More
Selleck Chemical LLC BAY 41-2272
SDP
Supplier:  Selleck Chemical LLC S81285MG
Encompass_Preferred
BAY 41-2272 is a direct and NO-independent soluble guanylate cyclase (sGC) stimulator
Catalog No. 50-000-00277