Learn More
Medchemexpress LLC HY-100029A 5mg Medchemexpress, Bay 41-4109 (racemate) CAS:298708-79-9 Purity:>98%
SDP
Catalog No. 501931699
Encompass_Preferred
Medchemexpress, HY-100029A 5mg Bay 41-4109 (racemate) CAS:298708-79-9 BAY 41-4109 racemate is the racemate of BAY 41-4109. BAY 41-4109 is a potent inhibitor of human hepatitis B virus ( HBV ) with an IC 50 of 53 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1699 Supplier Medchemexpress LLC Supplier No. HY100029A5MG
Medchemexpress, HY-100029A 5mg Bay 41-4109 (racemate) CAS:298708-79-9 BAY 41-4109 racemate is the racemate of BAY 41-4109. BAY 41-4109 is a potent inhibitor of human hepatitis B virus ( HBV ) with an IC 50 of 53 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.