Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bay 41-8543 | 256498-66-5 | MFCD18382081 | 99.9% | 420.44 g/mol | C21H21FN8O | 10 MG
SDP

Catalog No. 5000210553
Encompass_Preferred

BAY 41-8543 is a nitric oxide-independent, heme-dependent stimulator of soluble guanylyl cyclase (sGC) used as a research compound with vasodilator and antiplatelet activity. It is orally active in animal studies and is used for studies of cardiovascular and vascular biology.

  • Cas number: 256498-66-5.
  • Chemical formula: C21H21FN8O.
  • Molecular weight: 420.44 g/mol.
  • Purity: 99.9%.
  • Appearance: white to off-white solid.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Solubility: DMF 16.67 mg/mL, ethanol 1 mg/mL, DMSO <1 mg/mL.

Catalog No. 50-002-10553 Supplier Medchemexpress LLC Supplier No. HYW06283610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BAY 41-8543 is a nitric oxide-independent, heme-dependent stimulator of soluble guanylyl cyclase (sGC) used as a research compound with vasodilator and antiplatelet activity. It is orally active in animal studies and is used for studies of cardiovascular and vascular biology.

  • Cas number: 256498-66-5.
  • Chemical formula: C21H21FN8O.
  • Molecular weight: 420.44 g/mol.
  • Purity: 99.9%.
  • Appearance: white to off-white solid.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Solubility: DMF 16.67 mg/mL, ethanol 1 mg/mL, DMSO <1 mg/mL.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.