Learn More
Cayman Chemical BAY 412272 Nitric Oxide Reagen
SDP
Catalog No. 502889684
Encompass_Preferred
An activator of sGC, stimulating activity at concentrations as low as 10-100 nM; inhibits platelet aggregation (IC50 = 36 nM), induces relaxation of preconstricted aorta rings (IC50 = 304 nM); effective in vivo, decreasing mean arterial blood pressure in hypertensive rats
Catalog No. 50-288-9684 Supplier Cayman Chemical Supplier No. 1005750 MG
An activator of sGC, stimulating activity at concentrations as low as 10-100 nM; inhibits platelet aggregation (IC50 = 36 nM), induces relaxation of preconstricted aorta rings (IC50 = 304 nM); effective in vivo, decreasing mean arterial blood pressure in hypertensive rats