Learn More
Enzo Life Sciences BAY 43-9006 (1mg). CAS: 284461-73-0
SDP
Supplier:  Enzo Life Sciences ALX270469M001
Encompass_Preferred
Broad-spectrum kinase inhibitor. Alternative name: N-[4-Chloro-3-(trifluoromethyl)phenyl]-([4-[2-(N-methylcarbamoyl)(4-pyridyloxy)]phenyl]amino)-carboxamide, Sorafenib. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO, 100% ethanol, methanol or ethyl acetate. Long Term Storage: -20°C.
Catalog No. 50-200-8594