Learn More
Medchemexpress LLC BAY-524 | 1445830-39-6 | 98.4% | 482.48 g·mol⁻¹ | C24H24F2N6O3 | 1 ML
SDP
Catalog No. 5000244169
Encompass_Preferred
BAY-524 is a small-molecule Bub1 kinase inhibitor supplied as a solid or as a 10 mM solution in DMSO for research use. It has molecular formula C24H24F2N6O3, molecular weight 482.48 g·mol⁻¹, and a reported purity of 98.36%. The product is provided with solubility and storage guidance for in vitro and in vivo applications.
Catalog No. 50-002-44169 Supplier Medchemexpress LLC Supplier No. HY10400110MM 1ML
BAY-524 is a small-molecule Bub1 kinase inhibitor supplied as a solid or as a 10 mM solution in DMSO for research use. It has molecular formula C24H24F2N6O3, molecular weight 482.48 g·mol⁻¹, and a reported purity of 98.36%. The product is provided with solubility and storage guidance for in vitro and in vivo applications.