Learn More
Cayman Chemical BAY 55-9837 (trifluoroacetate salt), Purity: ≥98%, 5 mg
SDP
Catalog No. 502910019
Encompass_Preferred
A VPAC2 agonist; binds to VPAC2 (Kd = 65 nM) and selectively induces cAMP production in CHO cells expressing human VPAC2 over VPAC1 or PAC1 receptor (EC50s = 0.4, 100, and >1,000 nM, respectively); increases glucose-dependent insulin secretion in isolated rat and human pancreatic islets at 100 nM; enhances glucose-induced insulin secretion in fasted rats (ED50 = ~3 pmol/kg)
Catalog No. 50-291-0019 Supplier Cayman Chemical Supplier No. 367525 MG
A VPAC2 agonist; binds to VPAC2 (Kd = 65 nM) and selectively induces cAMP production in CHO cells expressing human VPAC2 over VPAC1 or PAC1 receptor (EC50s = 0.4, 100, and >1,000 nM, respectively); increases glucose-dependent insulin secretion in isolated rat and human pancreatic islets at 100 nM; enhances glucose-induced insulin secretion in fasted rats (ED50 = ~3 pmol/kg)