Learn More
Medchemexpress LLC HY-100488 5mg Medchemexpress, Bay 59-3074 CAS:406205-74-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1004885MG
Encompass_Preferred
Medchemexpress, HY-100488 5mg Bay 59-3074 CAS:406205-74-1 Bay 59-3074 is a selective cannabinoid CB 1 /CB 2 receptor partial agonist with K i values of 48.3 and 45.5 nM at human CB 1 and CB 2 receptors, respectively. Bay 59-3074 has analgesic properties [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1851