Learn More
Medchemexpress LLC Bay 59-9435 | 654059-21-9 | 99.8% | 266.34 g·mol⁻¹ | C14H22N2O3 | 25 MG
SDP
Catalog No. 5000245496
Encompass_Preferred
BAY 59-9435 is a potent, selective small-molecule inhibitor of hormone-sensitive lipase (HSL) with a reported IC50 of 0.023 μM. It is used as a research tool to study HSL-dependent lipolysis in cellular and animal models. Not for human or clinical use.
  • Potent, selective inhibition of hormone-sensitive lipase.
  • Low IC50 (0.023 μM) enabling submicromolar activity.
  • High purity suitable for biochemical and cellular assays (99.8%).
  • Characterized chemical profile (C14H22N2O3; MW 266.34 g·mol⁻¹).
  • Convenient small pack size for preclinical research (25 mg).
Catalog No. 50-002-45496 Supplier Medchemexpress LLC Supplier No. HY10205625MG
BAY 59-9435 is a potent, selective small-molecule inhibitor of hormone-sensitive lipase (HSL) with a reported IC50 of 0.023 μM. It is used as a research tool to study HSL-dependent lipolysis in cellular and animal models. Not for human or clinical use.
  • Potent, selective inhibition of hormone-sensitive lipase.
  • Low IC50 (0.023 μM) enabling submicromolar activity.
  • High purity suitable for biochemical and cellular assays (99.8%).
  • Characterized chemical profile (C14H22N2O3; MW 266.34 g·mol⁻¹).
  • Convenient small pack size for preclinical research (25 mg).