Learn More
Medchemexpress LLC Bay-593 | 2413020-56-9 | 99.9% | 476.53 | 1 ML
SDP
Catalog No. 5000394560
Encompass_Preferred
BAY-593 is an orally active GGTase-I inhibitor. It can block YAP1/TAZ signaling in animals and has antitumor activity. It is for research use only and not sold to patients.
  • Orally active GGTase-I inhibitor
  • Blocks YAP1/TAZ signaling in animals
  • Exhibits antitumor activity
  • Appearance: Solid
  • Color: White to off-white
Catalog No. 50-003-94560 Supplier Medchemexpress LLC Supplier No. HY16157310MM/1ML
BAY-593 is an orally active GGTase-I inhibitor. It can block YAP1/TAZ signaling in animals and has antitumor activity. It is for research use only and not sold to patients.
  • Orally active GGTase-I inhibitor
  • Blocks YAP1/TAZ signaling in animals
  • Exhibits antitumor activity
  • Appearance: Solid
  • Color: White to off-white