Learn More
Medchemexpress LLC Bay-593 | 2413020-56-9 | 99.9% | 476.53 g/mol | C26H31F3N2O3 | 10 MG
SDP
Catalog No. 5000288606
Encompass_Preferred
BAY-593 is an orally active geranylgeranyltransferase-I (GGTase-I) inhibitor that blocks YAP1/TAZ signaling and demonstrates antitumor activity in animal models. It is used as a research probe to study Rho-GTPase prenylation, YAP1/TAZ-dependent pathways, and related cancer biology applications.
  • Orally active GGTase-I inhibitor suitable for in vivo studies.
  • Blocks YAP1/TAZ signaling in cells and animals.
  • Demonstrates antitumor activity in animal models.
  • High purity solid form for research applications.
  • Available in small research pack sizes and as a DMSO solution for screening.
Catalog No. 50-002-88606 Supplier Medchemexpress LLC Supplier No. HY16157310MG
BAY-593 is an orally active geranylgeranyltransferase-I (GGTase-I) inhibitor that blocks YAP1/TAZ signaling and demonstrates antitumor activity in animal models. It is used as a research probe to study Rho-GTPase prenylation, YAP1/TAZ-dependent pathways, and related cancer biology applications.
  • Orally active GGTase-I inhibitor suitable for in vivo studies.
  • Blocks YAP1/TAZ signaling in cells and animals.
  • Demonstrates antitumor activity in animal models.
  • High purity solid form for research applications.
  • Available in small research pack sizes and as a DMSO solution for screening.