Learn More
Selleck Chemical LLC BAY-593 E1976-25MG
SDP
Catalog No. 5000040248
Encompass_Preferred
BAY-593 is an in vivo probe and small molecule inhibitor of geranylgeranyltransferase-I (GGTase-I) blocking oncogenic YAP1/TAZ It exhibits potent anti-tumor activity and inhibits cancer cell proliferation in vitro
Catalog No. 50-000-40248 Supplier Selleck Chemical LLC Supplier No. E197625MG
BAY-593 is an in vivo probe and small molecule inhibitor of geranylgeranyltransferase-I (GGTase-I) blocking oncogenic YAP1/TAZ It exhibits potent anti-tumor activity and inhibits cancer cell proliferation in vitro