Learn More
TARGETMOL CHEMICALS INC BAY-598 10MG
SDP
Catalog No. 502877788
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. BAY 598 is a potent and selective competitive inhibitor of SMYD2 lysine methyltransferase, IC50 values are 27 and 58 nM for biochemical and cellular activity assays, respectively. Purity 99.18%
Catalog No. 50-287-7788 Supplier TARGETMOL CHEMICALS INC Supplier No. T740310MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. BAY 598 is a potent and selective competitive inhibitor of SMYD2 lysine methyltransferase, IC50 values are 27 and 58 nM for biochemical and cellular activity assays, respectively. Purity 99.18%