Learn More
Medchemexpress LLC BAY-598 | 1906919-67-2 | 98.2% | 525.34 | 1 MG
SDP
Catalog No. 5000403275
Encompass_Preferred
BAY-598 is a chemical probe and a selective small molecule inhibitor of SMYD2 with an IC50 of 27 nM. It is intended for research use only. This inhibitor effectively blocks in vitro methylation of MAPKAPK3 by SMYD2 and reduces the growth of Kras;p53 mutant PDAC cells.
  • Selective small molecule inhibitor of SMYD2.
  • Inhibits in vitro methylation of MAPKAPK3 by SMYD2.
  • Reduces the growth of Kras;p53 mutant PDAC cells.
Catalog No. 50-004-03275 Supplier Medchemexpress LLC Supplier No. HY195461MG
BAY-598 is a chemical probe and a selective small molecule inhibitor of SMYD2 with an IC50 of 27 nM. It is intended for research use only. This inhibitor effectively blocks in vitro methylation of MAPKAPK3 by SMYD2 and reduces the growth of Kras;p53 mutant PDAC cells.
  • Selective small molecule inhibitor of SMYD2.
  • Inhibits in vitro methylation of MAPKAPK3 by SMYD2.
  • Reduces the growth of Kras;p53 mutant PDAC cells.