Learn More
Medchemexpress LLC BAY-598 | 1906919-67-2 | 99.33% | 525.34 | 1 ML
SDP
Catalog No. 5000400447
Encompass_Preferred
BAY-598 is a selective small molecule inhibitor of SMYD2, functioning as a chemical probe. It effectively blocks in vitro methylation of MAPKAPK3 by SMYD2 and reduces the growth of Kras;p53 mutant PDAC cells.
  • Selective SMYD2 inhibitor with an IC50 of 27 nM.
  • Blocks in vitro methylation of MAPKAPK3 by SMYD2.
  • Reduces growth of Kras;p53 mutant PDAC cells.
  • Suitable for research use only.
Catalog No. 50-004-00447 Supplier Medchemexpress LLC Supplier No. HY1954610MM/1ML
BAY-598 is a selective small molecule inhibitor of SMYD2, functioning as a chemical probe. It effectively blocks in vitro methylation of MAPKAPK3 by SMYD2 and reduces the growth of Kras;p53 mutant PDAC cells.
  • Selective SMYD2 inhibitor with an IC50 of 27 nM.
  • Blocks in vitro methylation of MAPKAPK3 by SMYD2.
  • Reduces growth of Kras;p53 mutant PDAC cells.
  • Suitable for research use only.