Learn More
Apexbio Technology LLC BAY 60-6583 910487-58-0 25mg
SDP
Supplier:  Apexbio Technology LLC B561225
Encompass_Preferred
BAY 60-6583 is a potent and selective agonist targeting the adenosine A2B receptor a G-protein-coupled receptor activated by elevated extracellular adenosine It is designed to activate the A2B receptor thereby stimulating cAMP production and modulating intracellular signaling pathways BAY 60-6583 exerts its biological activity primarily through promotion of cAMP-mediated signaling influencing gene transcription associated with glucocorticoid-response and cAMP-response elements In various cell-based assays BAY 60-6583 activates A2B receptor-dependent responses promoting the expression of anti-inflammatory genes such as CD200 CRISPLD2 and SOCS3 Based on these pharmacological properties BAY 60-6583 holds research potential in cardiovascular protection and inflammation-focused studies
Catalog No. 50-000-90804