Learn More
Medchemexpress LLC Bay 60-7550 | 439083-90-6 | 98.4% | 476.57 | 1 ML
SDP
Catalog No. 5000360064
Encompass_Preferred
Bay 60-7550 is a potent and selective inhibitor of phosphodiesterase 2 (PDE2), characterized by a Ki value of 3.8 nM. It has demonstrated anxiolytic effects by increasing cGMP signaling and shows selectivity for PDE2 over other PDE families. This compound is suitable for research applications, contributing to the understanding of various biological processes.
  • Potent and selective PDE2 inhibition.
  • Demonstrates anxiolytic properties through elevated cGMP signaling.
  • Exhibits significant selectivity over other PDE families.
  • Useful for in vitro studies to increase cGMP in neuronal cultures and reduce proliferation of PASMCs.
  • Effective in in vivo models for reversing stress-induced behavioral alterations and increasing exploratory behavior.
Catalog No. 50-003-60064 Supplier Medchemexpress LLC Supplier No. HY1499210MM/1ML
Bay 60-7550 is a potent and selective inhibitor of phosphodiesterase 2 (PDE2), characterized by a Ki value of 3.8 nM. It has demonstrated anxiolytic effects by increasing cGMP signaling and shows selectivity for PDE2 over other PDE families. This compound is suitable for research applications, contributing to the understanding of various biological processes.
  • Potent and selective PDE2 inhibition.
  • Demonstrates anxiolytic properties through elevated cGMP signaling.
  • Exhibits significant selectivity over other PDE families.
  • Useful for in vitro studies to increase cGMP in neuronal cultures and reduce proliferation of PASMCs.
  • Effective in in vivo models for reversing stress-induced behavioral alterations and increasing exploratory behavior.