Learn More
Medchemexpress LLC BAY-6035 | 2247890-13-5 | 99.51% | 396.48 | 1 ML
SDP
Catalog No. 5000436274
Encompass_Preferred
BAY-6035 is a chemical probe and a potent, selective, and substrate-competitive inhibitor of SMYD3. It inhibits the methylation of MEKK2 peptide with an IC50 of 88 nM.
  • Potent, selective, and substrate-competitive inhibitor of SMYD3.
  • Inhibits methylation of MEKK2 peptide with an IC50 of 88 nM.
  • Exhibits more than 100-fold selectivity over other histone methyltransferases.
  • Inhibits the methylation of MEKK2 in cells with an IC50 of 70 nM.
Catalog No. 50-004-36274 Supplier Medchemexpress LLC Supplier No. HY11208010MM/1ML
Add to Cart
Add to Cart
BAY-6035 is a chemical probe and a potent, selective, and substrate-competitive inhibitor of SMYD3. It inhibits the methylation of MEKK2 peptide with an IC50 of 88 nM.
  • Potent, selective, and substrate-competitive inhibitor of SMYD3.
  • Inhibits methylation of MEKK2 peptide with an IC50 of 88 nM.
  • Exhibits more than 100-fold selectivity over other histone methyltransferases.
  • Inhibits the methylation of MEKK2 in cells with an IC50 of 70 nM.