Learn More
Medchemexpress LLC BAY-6035 | 2247890-13-5 | 99.51% | 396.48 | 500 MG
SDP
Catalog No. 5000437159
Encompass_Preferred
BAY-6035 is a chemical probe that acts as a potent, selective, and substrate-competitive inhibitor of SMYD3. It inhibits the methylation of MEKK2 peptide with an IC50 of 88 nM.
  • Potent, selective, and substrate-competitive inhibitor of SMYD3
  • Inhibits methylation of MEKK2 peptide
  • Shows more than 100-fold selectivity over other histone methyltransferases
  • Inhibits the methylation of MEKK2 in cells
Catalog No. 50-004-37159 Supplier Medchemexpress LLC Supplier No. HY112080500MG
Add to Cart
Add to Cart
BAY-6035 is a chemical probe that acts as a potent, selective, and substrate-competitive inhibitor of SMYD3. It inhibits the methylation of MEKK2 peptide with an IC50 of 88 nM.
  • Potent, selective, and substrate-competitive inhibitor of SMYD3
  • Inhibits methylation of MEKK2 peptide
  • Shows more than 100-fold selectivity over other histone methyltransferases
  • Inhibits the methylation of MEKK2 in cells