Learn More
TARGETMOL CHEMICALS INC BAY 61-3606 5MG
SDP
Catalog No. 502579916
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BAY 61-3606 (Syk inhibitor IV) is a potent ATP-competitive reversible and highly selective inhibitor of Syk tyrosine kinase activity (Ki= 7.5 nM) exhibiting no inhibitory effect against Btk Fyn Itk Lyn and Src. purity: 98%
Catalog No. 50-257-9916 Supplier TARGETMOL CHEMICALS INC Supplier No. T42635MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BAY 61-3606 (Syk inhibitor IV) is a potent ATP-competitive reversible and highly selective inhibitor of Syk tyrosine kinase activity (Ki= 7.5 nM) exhibiting no inhibitory effect against Btk Fyn Itk Lyn and Src. purity: 98%