Learn More
Apexbio Technology LLC BAY 61-3606 732983-37-8 100mg
SDP
Catalog No. 5000095797
Encompass_Preferred
BAY 61-3606 (CAS 732983-37-8) is a selective orally bioavailable inhibitor of spleen tyrosine kinase (SYK) displaying an IC50 of 10 nM and a Ki of 7 5 nM for SYK by competitively binding ATP This compound shows minimal activity against other tyrosine kinases such as Src Fyn Lyn Btk and Itk at concentrations up to 4 7 M BAY 61-3606 suppresses Fc RI-mediated degranulation and mediator release in mast cells and basophils as well as Fc R-driven responses in macrophages and neutrophils In preclinical models BAY 61-3606 attenuates inflammatory cell functions and reduces allergic and asthmatic responses supporting its utility in investigating SYK-mediated signaling and inflammation
Catalog No. 50-000-95797 Supplier Apexbio Technology LLC Supplier No. A3227100
BAY 61-3606 (CAS 732983-37-8) is a selective orally bioavailable inhibitor of spleen tyrosine kinase (SYK) displaying an IC50 of 10 nM and a Ki of 7 5 nM for SYK by competitively binding ATP This compound shows minimal activity against other tyrosine kinases such as Src Fyn Lyn Btk and Itk at concentrations up to 4 7 M BAY 61-3606 suppresses Fc RI-mediated degranulation and mediator release in mast cells and basophils as well as Fc R-driven responses in macrophages and neutrophils In preclinical models BAY 61-3606 attenuates inflammatory cell functions and reduces allergic and asthmatic responses supporting its utility in investigating SYK-mediated signaling and inflammation