Learn More
Medchemexpress LLC Bay 61-3606 hydrochloride | 1615197-10-8 | MFCD29073361 | 98.1% | 426.86 | C20H19ClN6O3 | 10 MG
SDP
Catalog No. 5000299153
Encompass_Preferred
BAY 61-3606 hydrochloride is the hydrochloride salt of BAY 61-3606, a selective ATP-competitive Syk inhibitor used for in vitro research on Syk-mediated signaling and apoptosis. The product is supplied as a solid with a reported CAS number 1615197-10-8, molecular formula C20H19ClN6O3, molecular weight 426.86, and high purity for research applications.
  • Selective Syk inhibitor suitable for cell signaling studies.
  • ATP-competitive and reversible mechanism of action.
  • Hydrochloride salt form improves solubility for aqueous assays.
  • High reported purity supports reproducible experimental results.
  • Intended for research use only; not for human or veterinary use.
Catalog No. 50-002-99153 Supplier Medchemexpress LLC Supplier No. HY76474A10MG
BAY 61-3606 hydrochloride is the hydrochloride salt of BAY 61-3606, a selective ATP-competitive Syk inhibitor used for in vitro research on Syk-mediated signaling and apoptosis. The product is supplied as a solid with a reported CAS number 1615197-10-8, molecular formula C20H19ClN6O3, molecular weight 426.86, and high purity for research applications.
  • Selective Syk inhibitor suitable for cell signaling studies.
  • ATP-competitive and reversible mechanism of action.
  • Hydrochloride salt form improves solubility for aqueous assays.
  • High reported purity supports reproducible experimental results.
  • Intended for research use only; not for human or veterinary use.