Learn More
Medchemexpress LLC Bay 65-1942 hydrochloride | 600734-06-3 | 99.4% | C22H26ClN3O4 | 1 ML
SDP
Catalog No. 5000358395
Encompass_Preferred
Bay 65-1942 hydrochloride is an ATP-competitive and selective IKKβ inhibitor. It has been shown to significantly decrease left ventricular infarct size and attenuate CK-MB levels in research studies. In vitro, it can synergistically inhibit cell viability and induce caspase 3/7 activation.
  • ATP-competitive and selective IKKβ inhibitor
  • Decreases left ventricular infarct size
  • Attenuates CK-MB levels
  • Synergistically inhibits cell viability with AZD6244
  • Induces caspase 3/7 activation
Catalog No. 50-003-58395 Supplier Medchemexpress LLC Supplier No. HY5094810MM/1ML
Bay 65-1942 hydrochloride is an ATP-competitive and selective IKKβ inhibitor. It has been shown to significantly decrease left ventricular infarct size and attenuate CK-MB levels in research studies. In vitro, it can synergistically inhibit cell viability and induce caspase 3/7 activation.
  • ATP-competitive and selective IKKβ inhibitor
  • Decreases left ventricular infarct size
  • Attenuates CK-MB levels
  • Synergistically inhibits cell viability with AZD6244
  • Induces caspase 3/7 activation