Learn More
Medchemexpress LLC Bay 65-1942 hydrochloride | 600734-06-3 | 99.4% | C22H26ClN3O4 | 100 MG
SDP
Catalog No. 5000357784
Encompass_Preferred
Bay 65-1942 hydrochloride is an ATP-competitive and selective IKKβ inhibitor. This compound is intended for research use only. In vitro studies show that delivery of Bay 65-1942 prior to ischemia significantly decreases left ventricular infarct size. In vivo, it demonstrates synergistic inhibition of cell viability and induces caspase 3/7 activation when combined with AZD6244 in MYL-R cells.
  • Acts as an ATP-competitive and selective IKKβ inhibitor
  • Decreases left ventricular infarct size in vitro
  • Attenuates CK-MB levels in vivo
  • Demonstrates synergistic inhibition of cell viability with AZD6244
  • Induces caspase 3/7 activation with AZD6244
  • Suitable for research applications
Catalog No. 50-003-57784 Supplier Medchemexpress LLC Supplier No. HY50948100MG
Bay 65-1942 hydrochloride is an ATP-competitive and selective IKKβ inhibitor. This compound is intended for research use only. In vitro studies show that delivery of Bay 65-1942 prior to ischemia significantly decreases left ventricular infarct size. In vivo, it demonstrates synergistic inhibition of cell viability and induces caspase 3/7 activation when combined with AZD6244 in MYL-R cells.
  • Acts as an ATP-competitive and selective IKKβ inhibitor
  • Decreases left ventricular infarct size in vitro
  • Attenuates CK-MB levels in vivo
  • Demonstrates synergistic inhibition of cell viability with AZD6244
  • Induces caspase 3/7 activation with AZD6244
  • Suitable for research applications