Learn More
Medchemexpress LLC Bay 65-1942 hydrochloride | 600734-06-3 | 99.4% | C22H26ClN3O4 | 25 MG
SDP
Catalog No. 5000359010
Encompass_Preferred
Bay 65-1942 hydrochloride is an ATP-competitive and selective IKKβ inhibitor, appearing as a white to off-white solid. It has demonstrated efficacy in reducing left ventricular infarct size and attenuating CK-MB levels in vitro. In vivo studies show synergistic inhibition of cell viability when combined with AZD6244, and it induces caspase 3/7 activation, making it a valuable tool for research in cancer, inflammation, and immune system diseases.
  • ATP-competitive and selective IKKβ inhibitor
  • White to off-white solid appearance
  • Soluble in DMSO (50 mg/mL) and H2O (2.17 mg/mL)
  • Inhibits IKKβ activity
  • Decreases left ventricular infarct size in vitro
  • Attenuates CK-MB levels in vitro
  • Demonstrates synergistic inhibition of cell viability with AZD6244 in vivo
  • Induces caspase 3/7 activation in vivo
  • Relevant for research in cancer, inflammation, and immune system diseases
Catalog No. 50-003-59010 Supplier Medchemexpress LLC Supplier No. 25MGHY50948
Bay 65-1942 hydrochloride is an ATP-competitive and selective IKKβ inhibitor, appearing as a white to off-white solid. It has demonstrated efficacy in reducing left ventricular infarct size and attenuating CK-MB levels in vitro. In vivo studies show synergistic inhibition of cell viability when combined with AZD6244, and it induces caspase 3/7 activation, making it a valuable tool for research in cancer, inflammation, and immune system diseases.
  • ATP-competitive and selective IKKβ inhibitor
  • White to off-white solid appearance
  • Soluble in DMSO (50 mg/mL) and H2O (2.17 mg/mL)
  • Inhibits IKKβ activity
  • Decreases left ventricular infarct size in vitro
  • Attenuates CK-MB levels in vitro
  • Demonstrates synergistic inhibition of cell viability with AZD6244 in vivo
  • Induces caspase 3/7 activation in vivo
  • Relevant for research in cancer, inflammation, and immune system diseases