Learn More
Medchemexpress LLC Bay 65-1942 hydrochloride | 600734-06-3 | MFCD11040976 | 99.4% | 431.91 g/mol | C22H26ClN3O4 | 10 MG
SDP
Catalog No. 5000298795
Encompass_Preferred
Bay 65-1942 hydrochloride is an ATP-competitive, selective inhibitor of IKKβ used in research to probe NF-κB signaling and inflammatory pathways. It is supplied as the hydrochloride salt to improve solubility and handling, and is intended for use in biochemical and cell-based assays.
  • Selective inhibition of IKKβ for pathway interrogation.
  • ATP-competitive mode of action suitable for biochemical assays.
  • High reported purity for reproducible results.
  • Hydrochloride salt with improved solubility in DMSO and aqueous buffers.
  • Suitable for biochemical and cell-based assay applications.
Catalog No. 50-002-98795 Supplier Medchemexpress LLC Supplier No. HY5094810MG
Bay 65-1942 hydrochloride is an ATP-competitive, selective inhibitor of IKKβ used in research to probe NF-κB signaling and inflammatory pathways. It is supplied as the hydrochloride salt to improve solubility and handling, and is intended for use in biochemical and cell-based assays.
  • Selective inhibition of IKKβ for pathway interrogation.
  • ATP-competitive mode of action suitable for biochemical assays.
  • High reported purity for reproducible results.
  • Hydrochloride salt with improved solubility in DMSO and aqueous buffers.
  • Suitable for biochemical and cell-based assay applications.