Learn More
Medchemexpress LLC Bay-6672 hydrochloride | 2247520-31-4 | 98.0% | 581.33 | C26H28BrCl2N3O3 | 5 MG
SDP
Catalog No. 5000220994
Encompass_Preferred
BAY-6672 hydrochloride is a research-grade small molecule antagonist of the human Prostaglandin F (FP) receptor with a reported IC50 of 11 nM. Supplied as the hydrochloride salt and characterized at high purity, it is intended for in vitro pharmacology and biochemical studies and is available as a solid or as a 10 mM solution in DMSO. For research use only; not for human or clinical use.
  • Potent FP receptor antagonist (IC50 = 11 nM).
  • High purity suitable for pharmacological assays.
  • Available as solid mg quantities and as a 10 mM DMSO solution.
  • Characterized molecular weight and formula for identification.
  • Intended for in vitro research use only.
Catalog No. 50-002-20994 Supplier Medchemexpress LLC Supplier No. HY142658A5MG
BAY-6672 hydrochloride is a research-grade small molecule antagonist of the human Prostaglandin F (FP) receptor with a reported IC50 of 11 nM. Supplied as the hydrochloride salt and characterized at high purity, it is intended for in vitro pharmacology and biochemical studies and is available as a solid or as a 10 mM solution in DMSO. For research use only; not for human or clinical use.
  • Potent FP receptor antagonist (IC50 = 11 nM).
  • High purity suitable for pharmacological assays.
  • Available as solid mg quantities and as a 10 mM DMSO solution.
  • Characterized molecular weight and formula for identification.
  • Intended for in vitro research use only.