Learn More
Medchemexpress LLC Bay-6672 hydrochloride | 2247520-31-4 | 98.0% | C26H28BrCl2N3O3 | 10MG
SDP
Catalog No. 5000261306
Encompass_Preferred
BAY-6672 hydrochloride is a potent, selective antagonist of the human Prostaglandin F (FP) receptor (reported IC50 ≈ 11 nM). It is used as a research reagent to probe FP receptor signaling and pharmacology and has demonstrated in vivo efficacy in preclinical lung fibrosis models. The compound is supplied as a solid with high reported purity.
  • Potent FP receptor antagonism (IC50 ≈ 11 nM).
  • Demonstrated in vivo efficacy in preclinical lung fibrosis models.
  • Selective for FP over related prostaglandin receptors in published assays.
  • High reported purity (~98.0%), suitable for biochemical studies.
  • Provided as a solid research reagent for pharmacology and mechanistic assays.
Catalog No. 50-002-61306 Supplier Medchemexpress LLC Supplier No. HY142658A10MG
BAY-6672 hydrochloride is a potent, selective antagonist of the human Prostaglandin F (FP) receptor (reported IC50 ≈ 11 nM). It is used as a research reagent to probe FP receptor signaling and pharmacology and has demonstrated in vivo efficacy in preclinical lung fibrosis models. The compound is supplied as a solid with high reported purity.
  • Potent FP receptor antagonism (IC50 ≈ 11 nM).
  • Demonstrated in vivo efficacy in preclinical lung fibrosis models.
  • Selective for FP over related prostaglandin receptors in published assays.
  • High reported purity (~98.0%), suitable for biochemical studies.
  • Provided as a solid research reagent for pharmacology and mechanistic assays.