Learn More
Medchemexpress LLC BAY-6672 hydrochloride | 2247520-31-4 | C26H28BrCl2N3O3 | 1 ML
SDP
Catalog No. 5000353583
Encompass_Preferred
BAY-6672 hydrochloride is a chemical compound supplied as a solid, primarily intended for laboratory research and the manufacture of substances.
  • Identified for laboratory chemical applications
  • Recommended storage at 4°C under nitrogen, away from moisture
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, under nitrogen
  • Ships at room temperature if transit is less than 2 weeks
  • For research use only
Catalog No. 50-003-53583 Supplier Medchemexpress LLC Supplier No. HY142658A10MM/1ML
BAY-6672 hydrochloride is a chemical compound supplied as a solid, primarily intended for laboratory research and the manufacture of substances.
  • Identified for laboratory chemical applications
  • Recommended storage at 4°C under nitrogen, away from moisture
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, under nitrogen
  • Ships at room temperature if transit is less than 2 weeks
  • For research use only