Learn More
Medchemexpress LLC BAY-677 | 2117404-84-7 | 98.0% | 400.35 | 10 MG
SDP
Catalog No. 5000345556
Encompass_Preferred
BAY-677 serves as an inactive control for BAY-678. BAY-678 is an orally bioavailable, highly potent, selective, and cell-permeable inhibitor of human neutrophil elastase (HNE), demonstrating an IC50 of 20 nM. This compound has also been nominated as a chemical probe to the public via the Structural Genomics Consortium (SGC).
  • Inactive control for BAY-678.
  • BAY-678 is an orally bioavailable inhibitor.
  • Highly potent and selective inhibitor of human neutrophil elastase.
  • Cell-permeable.
  • IC50 of 20 nM against HNE.
  • Nominated as a chemical probe by SGC.
Catalog No. 50-003-45556 Supplier Medchemexpress LLC Supplier No. HY11145750MG
BAY-677 serves as an inactive control for BAY-678. BAY-678 is an orally bioavailable, highly potent, selective, and cell-permeable inhibitor of human neutrophil elastase (HNE), demonstrating an IC50 of 20 nM. This compound has also been nominated as a chemical probe to the public via the Structural Genomics Consortium (SGC).
  • Inactive control for BAY-678.
  • BAY-678 is an orally bioavailable inhibitor.
  • Highly potent and selective inhibitor of human neutrophil elastase.
  • Cell-permeable.
  • IC50 of 20 nM against HNE.
  • Nominated as a chemical probe by SGC.