Learn More
Medchemexpress LLC BAY-677 | 2117404-84-7 | 98.0% | 400.35 | 50 MG
SDP
Catalog No. 5000345684
Encompass_Preferred
BAY-677 serves as an inactive control for BAY-678. BAY-678 is an orally bioavailable, highly potent, selective, and cell-permeable inhibitor of human neutrophil elastase (HNE). BAY-678 has also been designated as a chemical probe for public use through the Structural Genomics Consortium (SGC).
  • Inactive control for BAY-678
  • Used in research for human neutrophil elastase (HNE) studies
  • Related to a chemical probe designated for public use
Catalog No. 50-003-45684 Supplier Medchemexpress LLC Supplier No. HY111457100MG
BAY-677 serves as an inactive control for BAY-678. BAY-678 is an orally bioavailable, highly potent, selective, and cell-permeable inhibitor of human neutrophil elastase (HNE). BAY-678 has also been designated as a chemical probe for public use through the Structural Genomics Consortium (SGC).
  • Inactive control for BAY-678
  • Used in research for human neutrophil elastase (HNE) studies
  • Related to a chemical probe designated for public use