Learn More
Medchemexpress LLC HY-111457 5mg Medchemexpress, BAY-677 CAS:2117404-84-7 Purity:>98%
SDP
Catalog No. 501932365
Encompass_Preferred
Medchemexpress, HY-111457 5mg BAY-677 CAS:2117404-84-7 BAY-677 is an inactive control for BAY-678. BAY-678 is an orally bioavailable, highly potent, selective and cell-permeable inhibitor of human neutrophil elastase (HNE) , with an IC 50 of 20 nM [1] . BAY-678 is also nominated as a chemical probe to the public via the Structural Genomics Consortium (SGC) [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2365 Supplier Medchemexpress LLC Supplier No. HY1114575MG
Medchemexpress, HY-111457 5mg BAY-677 CAS:2117404-84-7 BAY-677 is an inactive control for BAY-678. BAY-678 is an orally bioavailable, highly potent, selective and cell-permeable inhibitor of human neutrophil elastase (HNE) , with an IC 50 of 20 nM [1] . BAY-678 is also nominated as a chemical probe to the public via the Structural Genomics Consortium (SGC) [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.