Learn More
Medchemexpress LLC HY-111457A 10mg Medchemexpress, BAY-678 CAS:675103-36-3 Purity:>98%
SDP
Catalog No. 501968876
Encompass_Preferred
Medchemexpress, HY-111457A 10mg BAY-678 CAS:675103-36-3 BAY-678 is an orally bioavailable, highly potent, selective and cell-permeable inhibitor of human neutrophil elastase (HNE), with an IC50 of 20 nM. BAY-678 is also nominated as a chemical probe to the public via the Structural Genomics Consortium (SGC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8876 Supplier Medchemexpress LLC Supplier No. HY111457A10MG
Medchemexpress, HY-111457A 10mg BAY-678 CAS:675103-36-3 BAY-678 is an orally bioavailable, highly potent, selective and cell-permeable inhibitor of human neutrophil elastase (HNE), with an IC50 of 20 nM. BAY-678 is also nominated as a chemical probe to the public via the Structural Genomics Consortium (SGC). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.